α-Bromo-2,6-difluorotoluene ≥97%

Danger

Synonyms: 2-(Bromomethyl)-1,3-difluorobenzene, 2,6-Difluorobenzyl bromide

264431-25G 264431-5G
220048-803EA 127.08 USD
220048-803 220048-804
α-Bromo-2,6-difluorotoluene ≥97%
α-Bromo-2,6-difluorotoluene
Formula: C₇H₅BrF₂
MW: 207.02 g/mol
Melting Pt: 53 °C
Density: 1.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000329
CAS Number: 85118-00-9
EINECS: 285-652-6
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR