α-Bromo-2,5-difluorotoluene ≥98%

Synonyms: 2-(Bromomethyl)-1,4-difluorobenzene, 2,5-Difluorobenzyl bromide

264423-1G 264423-10G
220048-802EA 35.08 USD
220048-802 220048-801
α-Bromo-2,5-difluorotoluene ≥98%
α-Bromo-2,5-difluorotoluene
Formula: C₇H₅BrF₂
MW: 207.02 g/mol
Boiling Pt: 186 °C (1013 hPa)
Melting Pt: 8 °C
Density: 1.609 g/cm³ (20 °C)
MDL Number: MFCD00009897
CAS Number: 85117-99-3
EINECS: 285-651-0
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR