3,4-Difluorobenzylamine ≥98%

Danger

264407-1G 264407-5G
220048-797EA 62.46 USD
220048-797 220048-798
3,4-Difluorobenzylamine ≥98%
3,4-Difluorobenzylamine
Formula: C₇H₇F₂N
MW: 143.14 g/mol
Boiling Pt: 174 °C (1013 hPa)
Melting Pt: 31 °C
Density: 1.21 g/cm³ (20 °C)
MDL Number: MFCD00010145
CAS Number: 72235-53-1
EINECS: 276-503-6
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR