2,6-Difluorobenzylamine ≥97%

Danger

264393-1G 264393-5G
220048-795EA 35.32 USD
220048-795 220048-796
2,6-Difluorobenzylamine ≥97%
2,6-Difluorobenzylamine
Formula: C₇H₇F₂N
MW: 143.14 g/mol
Boiling Pt: 174 - 177 °C (1013 hPa)
Density: 1.19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010144
CAS Number: 69385-30-4
EINECS: 273-983-9
UN: 2735
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR