2,5-Difluorobenzylamine ≥97%

Danger

Synonyms: (2,5-Difluorophenyl)methanamine

264385-5G 264385-1G
220048-794EA 75.8 USD
220048-794 220048-793
2,5-Difluorobenzylamine ≥97%
2,5-Difluorobenzylamine
Formula: C₇H₇F₂N
MW: 143.14 g/mol
Boiling Pt: 76…78 °C (15 mmHg)
Density: 1.22 g/cm³ (20 °C)
MDL Number: MFCD00010143
CAS Number: 85118-06-5
EINECS: 285-658-9
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR