2,4-Difluorobenzylamine ≥98%

Danger

Synonyms: (2,4-Difluorophenyl)methanamine

264377-1G 264377-5G
220048-791EA 35.65 USD
220048-791 220048-792
2,4-Difluorobenzylamine ≥98%
2,4-Difluorobenzylamine
Formula: C₇H₇F₂N
MW: 143.14 g/mol
Boiling Pt: 82…84 °C (15 mmHg)
Density: 1.2 g/cm³ (20 °C)
MDL Number: MFCD00010142
CAS Number: 72235-52-0
EINECS: 276-502-0
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR