2,4-Difluorobenzonitrile ≥97%

264326-25G 264326-5G
220048-788EA 58.05 USD
220048-788 220048-789
2,4-Difluorobenzonitrile ≥97%
2,4-Difluorobenzonitrile
Formula: C₇H₃F₂N
MW: 139.1 g/mol
Boiling Pt: 80…83 °C (22.5 mmHg)
Melting Pt: 46…47 °C
MDL Number: MFCD00009826
CAS Number: 3939-09-1
EINECS: 223-523-8
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR