2,4-Difluorobenzoic acid ≥98%

Warning

264296-25G 264296-5G
220048-782EA 82.28 USD
220048-782 220048-783
2,4-Difluorobenzoic acid ≥98%
2,4-Difluorobenzoic acid
Formula: C₇H₄F₂O₂
MW: 158.1 g/mol
Boiling Pt: 241 °C (1013 hPa)
Melting Pt: 185 °C
Storage Temperature: Ambient
MDL Number: MFCD00011670
CAS Number: 1583-58-0
EINECS: 216-430-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR