n-Tricosane ≥99%

Synonyms: Tricosane

263850-5G 263850-1G
220049-408EA 58.26 USD
220049-408 220049-407
n-Tricosane ≥99%
n-Tricosane
Formula: H₃C(CH₂)₂₁CH₃
MW: 324.63 g/mol
Boiling Pt: 380 °C (1013 hPa)
Melting Pt: 46…47.5 °C
Density: 0.7969 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009350
CAS Number: 638-67-5
EINECS: 211-347-4

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR