trans-4-Aminocyclohexanol hydrochloride ≥97%

263761-100G 263761-25G
220049-400EA 93.69 USD
220049-400 220049-401
trans-4-Aminocyclohexanol hydrochloride ≥97%
trans-4-Aminocyclohexanol hydrochloride
Formula: C₆H₁₄ClNO
MW: 151.64 g/mol
MDL Number: MFCD00012566
CAS Number: 50910-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR