1,4-Bis(trimethylsilyl)-1,3-butadiyne

Synonyms: 1,4-Bis(trimethylsilyl)buta-1,3-diyne, 1,4-Bis(trimethylsilyl)butadiyne

263567-5G 263567-25G
220049-385EA 202.46 USD
220049-385 220049-384
1,4-Bis(trimethylsilyl)-1,3-butadiyne
1,4-Bis(trimethylsilyl)-1,3-butadiyne
Formula: C₁₀H₁₈Si₂
MW: 194.42 g/mol
MDL Number: MFCD00009839
CAS Number: 4526-07-2
UN: 1325
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR