3,5-Difluoroaniline ≥98%

Warning

263532-5G 263532-1G
220048-959EA 65.77 USD
220048-959 220048-958
3,5-Difluoroaniline ≥98%
3,5-Difluoroaniline
Formula: C₆H₅F₂N
MW: 129.11 g/mol
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 37…41 °C
Density: 1.295 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007763
CAS Number: 372-39-4
EINECS: 206-752-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR