2,3-Difluorobenzoic acid ≥98%

Warning

263516-1G 263516-10G
220048-956EA 31.12 USD
220048-956 220048-955
2,3-Difluorobenzoic acid ≥98%
2,3-Difluorobenzoic acid
Formula: C₇H₄F₂O₂
MW: 158.1 g/mol
MDL Number: MFCD00010267
CAS Number: 4519-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR