4-(Trifluoromethyl)benzylamine ≥97%

263508-10G 263508-1G
220048-953EA 206.77 USD
220048-953 220048-954
4-(Trifluoromethyl)benzylamine ≥97%
4-(Trifluoromethyl)benzylamine
Formula: C₈H₈F₃N
MW: 175.15 g/mol
Boiling Pt: 95…97 °C (1013 hPa)
Density: 1.229 g/cm³ (20 °C)
MDL Number: MFCD00010220
CAS Number: 3300-51-4
EINECS: 221-971-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR