3-(Trifluoromethyl)benzylamine ≥98%

Danger

263494-25G 263494-5G
220048-951EA 221.85 USD
220048-951 220048-952
3-(Trifluoromethyl)benzylamine ≥98%
3-(Trifluoromethyl)benzylamine
Formula: C₈H₈F₃N
MW: 175.15 g/mol
Boiling Pt: 72…74 °C (13 mmHg)
Density: 1.22 g/cm³ (20 °C)
MDL Number: MFCD00008117
CAS Number: 2740-83-2
EINECS: 220-367-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR