1-Bromo-2,4-difluorobenzene ≥98%

Danger

Synonyms: 2,4-Difluorobromobenzene

263443-100G 263443-25G
220048-946EA 88.46 USD
220048-946 220048-947
1-Bromo-2,4-difluorobenzene ≥98%
1-Bromo-2,4-difluorobenzene
Formula: C₆H₃BrF₂
MW: 192.99 g/mol
Boiling Pt: 145 °C (1013 hPa)
Melting Pt: -4 °C
Density: 1.69 g/cm³ (20 °C)
Flash Pt: 51 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000330
CAS Number: 348-57-2
EINECS: 206-479-4
UN: 2924
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR