3,5-Bis(trifluoromethyl)benzyl bromide ≥97%

Synonyms: 1-(Bromomethyl)-3,5-bis(trifluoromethyl)benzene

263397-5G 263397-1G
220048-942EA 65.15 USD
220048-942 220049-382
3,5-Bis(trifluoromethyl)benzyl bromide ≥97%
3,5-Bis(trifluoromethyl)benzyl bromide
Formula: C₉H₅BrF₆
MW: 307.03 g/mol
Boiling Pt: 136…140 °C (14 mmHg)
Density: 1.67 g/cm³ (20 °C)
MDL Number: MFCD00009905
CAS Number: 32247-96-4
EINECS: 250-971-1
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR