3,5-Bis(trifluoromethyl)benzylamine ≥80%, Technical Grade

Synonyms: (3,5-Bis(trifluoromethyl)phenyl)methanamine

263389-5G 263389-1G
220049-381EA 34.46 USD
220049-381 220049-380
3,5-Bis(trifluoromethyl)benzylamine ≥80%, Technical Grade
3,5-Bis(trifluoromethyl)benzylamine
Formula: C₉H₇F₆N
MW: 243.15 g/mol
Boiling Pt: 82…84 °C (15 mmHg)
Melting Pt: 46…55 °C
MDL Number: MFCD00009909
CAS Number: 85068-29-7
EINECS: 285-290-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR