2-Bromo-5-methylpyridine

Warning

263354-25G 263354-5G
220049-376EA 426.46 USD
220049-376 220049-377
2-Bromo-5-methylpyridine
2-Bromo-5-methylpyridine
Formula: C₆H₆BrN
Boiling Pt: 65 °C (0.5 -rr)
Melting Pt: 152.5 °C
MDL Number: MFCD00209553
CAS Number: 3510-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR