Tantalum ≥99.9%

Warning

262846-100G 262846-25G
220049-334EA 846.15 USD
220049-334 220049-335
Tantalum ≥99.9%
Tantalum
Formula: Ta
Density: 16.6 g/cm³ (20 °C)
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-135-5
UN: 3089
ADR: 4.1,II

Order Now

Specification Test Results

Assay 99.9% trace metals basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR