1-Iodoperfluorooctane ≥98%

Warning

Synonyms: Perfluorooctyl iodide, Heptadecafluoro-1-iodooctane, Perfluoro-1-iodooctane

262544-5G 262544-25G
220048-879EA 55.78 USD
220048-879 220048-878
1-Iodoperfluorooctane ≥98%
1-Iodoperfluorooctane
Formula: C₈F₁₇I
MW: 545.97 g/mol
Boiling Pt: 160 °C (1013 hPa)
Melting Pt: 25 °C
Density: 2.071 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001064
CAS Number: 507-63-1
EINECS: 208-079-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR