Biphenyl-3-ol ≥85%

Warning

Synonyms: 3-Phenylphenol, [1,1'-Biphenyl]-3-ol, 3-Hydroxybiphenyl, 3-Hydroxydiphenyl

262250-25G 262250-5G
220049-288EA 637.54 USD
220049-288 220049-289
Biphenyl-3-ol ≥85%
Biphenyl-3-ol
Formula: C₁₂H₁₀O
MW: 170.21 g/mol
Boiling Pt: 334 °C (1013 hPa)
Melting Pt: 78 °C
MDL Number: MFCD00002294
CAS Number: 580-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR