Phosphoryl trichloride ≥99.999%

Danger

262099-100G 262099-50G 262099-10G
220048-115EA 720.46 USD
220048-115 220048-117 220048-116
Phosphoryl trichloride ≥99.999%
Phosphoryl trichloride
Formula: POCl₃
Boiling Pt: 107 °C (1013 hPa)
Melting Pt: 1 °C
Density: 1.645 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011443
CAS Number: 10025-87-3
EINECS: 233-046-7
UN: 1810
ADR: 8,I
Merck Index: 12,07506

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR