Lanthanum(III) chloride heptahydrate ACS, Sigma-Aldrich®

Danger

262072-100G 262072-25G 262072-500G
220048-829EA 120 USD
220048-829 220048-830 220048-114
Lanthanum(III) chloride heptahydrate ACS, Sigma-Aldrich®
Lanthanum(III) chloride heptahydrate
Formula: LaCl₃·7H₂O
MW: 371.37 g/mol
Boiling Pt: ? 860 °C (1013 hPa)
Melting Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00149756
CAS Number: 10025-84-0
EINECS: 233-237-5
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR