Tricyclohexylphosphine

Warning

261971-1G 261971-25G 261971-5G
220049-271EA 48.94 USD
220049-271 220049-272 220049-273
Tricyclohexylphosphine
Tricyclohexylphosphine
Formula: C₁₈H₃₃P
Boiling Pt: 132…134 °C (0.02 mmHg)
Melting Pt: 78 °C
Density: 0.9 g/cm³ (20 °C)
Storage Temperature: Freezer
MDL Number: MFCD00003853
CAS Number: 2622-14-2
EINECS: 220-069-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR