1,4-Bis(diphenylphosphino)butane

Synonyms: DPPB, 1,4-Bis(diphenylphosphino)butane, Butylenebis(diphenylphosphine), Bis(1,2-diphenylphosphino)butane, Butane-1,3-diylbis(diphenylphosphine)

261947-25G 261947-5G
220049-264EA 60.43 USD
220049-264 220049-266
1,4-Bis(diphenylphosphino)butane
1,4-Bis(diphenylphosphino)butane
Formula: C₂₈H₂₈P₂
MW: 426.48 g/mol
Melting Pt: 195 °C
Density: 1.23 g/cm³ (20 °C)
MDL Number: MFCD00003051
CAS Number: 7688-25-7
EINECS: 231-698-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR