4-Aminopyrimidine

Warning

261823-1G 261823-250MG
220048-450EA 117.38 USD
220048-450 220048-451
4-Aminopyrimidine
4-Aminopyrimidine
Formula: C₄H₅N₃
MW: 95.1 g/mol
Melting Pt: 154 - 156 °C
MDL Number: MFCD00006112
CAS Number: 591-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR