2-(4-Aminophenyl)ethanol ≥98%

Synonyms: 4-Aminophenethyl alcohol

261645-25G 261645-5G
220049-261EA 189.54 USD
220049-261 220047-831
2-(4-Aminophenyl)ethanol ≥98%
2-(4-Aminophenyl)ethanol
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 235…255 °C (1013 hPa)
Melting Pt: 108 °C
MDL Number: MFCD00007922
CAS Number: 104-10-9
EINECS: 203-174-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR