4-Amino-N,N-diethylaniline ≥97%

Danger

Synonyms: N,N-Diethyl-1,4-phenylenediamine, N,N-Diethyl-p-phenylenediamine, N,N-Diethylbenzene-1,4-diamine, N,N-Diethylbenzene-p-diamine, N-1,N-1-Diethyl-1,4-benzenediamine

261513-100G 261513-25G
220048-772EA 199.23 USD
220048-772 220048-773
4-Amino-N,N-diethylaniline ≥97%
4-Amino-N,N-diethylaniline
Formula: C₁₀H₁₆N₂
MW: 164.25 g/mol
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 23 °C
Density: 0.988 g/cm³ (20 °C)
Flash Pt: 139 °C
MDL Number: MFCD00007861
CAS Number: 93-05-0
EINECS: 202-214-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR