1,1-Dimethylurea ≥99%

Synonyms: N,N-Dimethylurea

261394-25G 261394-100G
220049-251EA 108.77 USD
220049-251 220049-250
1,1-Dimethylurea ≥99%
1,1-Dimethylurea
Formula: C₃H₈N₂O
MW: 88.11 g/mol
MDL Number: MFCD00007959
CAS Number: 598-94-7
EINECS: 209-957-0

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR