3-Chloro-1,1-diethoxypropane, Technical Grade

Synonyms: 3-Chloropropionaldehydediethylacetal, 3-Chloropropionaldehyde diethyl acetal

26130-100ML 26130-25ML
220049-243EA 207.69 USD
220049-243 220049-244
3-Chloro-1,1-diethoxypropane, Technical Grade
3-Chloro-1,1-diethoxypropane
Formula: C₇H₁₅ClO₂
MW: 166.65 g/mol
Boiling Pt: 84 °C (1013 hPa)
Density: 0.979 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000994
CAS Number: 35573-93-4
EINECS: 252-623-4
UN: 1989
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR