4-Bromocinnamic acid predominantly trans

Warning

Synonyms: 3-(4-Bromophenyl)acrylic acid

260975-25G 260975-5G
220049-221EA 64.08 USD
220049-221 220049-222
4-Bromocinnamic acid predominantly trans
4-Bromocinnamic acid
Formula: C₉H₇BrO₂
MW: 227.06 g/mol
MDL Number: MFCD00004394
CAS Number: 1200-07-3
EINECS: 214-850-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR