8-Aminoquinoline ≥98%

Warning

260789-25G 260789-5G
220048-587EA 222.92 USD
220048-587 220048-588
8-Aminoquinoline ≥98%
8-Aminoquinoline
Formula: C₉H₈N₂
Boiling Pt: 290 °C (1013 hPa)
Melting Pt: 64 °C
Density: 1.337 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006809
CAS Number: 578-66-5
EINECS: 209-427-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR