(±)-1,2-Pentanediol ≥96%

Synonyms: (±)-1,2-Dihydroxy pentane, 1,2-Pentanediol

260282-250ML 260282-50ML
220049-213EA 553.15 USD
220049-213 220049-214
(±)-1,2-Pentanediol ≥96%
(±)-1,2-Pentanediol
Formula: C₅H₁₂O₂
MW: 104.15 g/mol
Boiling Pt: 206 °C (1013 hPa)
Density: 0.9802 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010736
CAS Number: 5343-92-0
EINECS: 226-285-3

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR