(2-Bromophenyl)acetic acid ≥99%

Synonyms: 2-(2-Bromophenyl)acetic acid

260061-5G 260061-25G
220049-196EA 45.23 USD
220049-196 220049-195
(2-Bromophenyl)acetic acid ≥99%
(2-Bromophenyl)acetic acid
Formula: (C₆H₄Br)CH₂COOH
MW: 215.05 g/mol
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 107.5 °C
MDL Number: MFCD00004314
CAS Number: 18698-97-0
EINECS: 242-509-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR