2-Bromo-6-nitrotoluene ≥98%

Warning

260053-5G 260053-25G
220049-194EA 64.4 USD
220049-194 220049-193
2-Bromo-6-nitrotoluene ≥98%
2-Bromo-6-nitrotoluene
Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Boiling Pt: 143 °C (22 mmHg)
Melting Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00009792
CAS Number: 55289-35-5
EINECS: 259-566-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR