Itaconic anhydride

Warning

Synonyms: Methylenesuccinic anhydride, 3-Methylenedihydrofuran-2,5-dione

259926-100G 259926-25G
220049-189EA 219.69 USD
220049-189 220049-190
Itaconic anhydride
Itaconic anhydride
Formula: C₅H₄O₃
MW: 112.08 g/mol
Boiling Pt: 239 °C (1013 hPa)
Melting Pt: 64…65 °C
MDL Number: MFCD00005530
CAS Number: 2170-03-8
EINECS: 218-518-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR