Toluene-p-sulfonic anhydride ≥97%

Synonyms: 4-Methylbenzenesulphonic anhydride, p-Toluenesulphonic anhydride

259764-5G 259764-25G
220049-181EA 56.75 USD
220049-181 220049-180
Toluene-p-sulfonic anhydride ≥97%
Toluene-p-sulphonic anhydride
Formula: C₁₄H₁₄O₅S₂
MW: 326.39 g/mol
MDL Number: MFCD00008548
CAS Number: 4124-41-8
EINECS: 223-926-9
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR