3,4,5-Trihydroxybenzaldehyde monohydrate ≥98%

Warning

Synonyms: 3,4,5-Trihydroxybenzaldehyde hydrate

259594-1G 259594-5G
220049-164EA 160.46 USD
220049-164 220049-165
3,4,5-Trihydroxybenzaldehyde monohydrate ≥98%
3,4,5-Trihydroxybenzaldehyde monohydrate
Formula: C₇H₆O₄·1H₂O
MW: 172.14 g/mol
MDL Number: MFCD00149124
CAS Number: 207742-88-9
EINECS: 237-168-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR