(4-Chlorophenoxy)acetic acid ≥98.0% (by titrimetric analysis)

Warning

Synonyms: 4-CPA

25890-250G 25890-1KG
220048-758EA 86.48 USD
220048-758 220048-757
(4-Chlorophenoxy)acetic acid ≥98.0% (by titrimetric analysis)
(4-Chlorophenoxy)acetic acid
Formula: (C₆H₄Cl)OCH₂COOH
MW: 186.59 g/mol
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 160 °C
Density: 1.53 g/cm³ (20 °C)
MDL Number: MFCD00004305
CAS Number: 122-88-3
EINECS: 204-581-3

Order Now

Specification Test Results

Assay 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR