1-Hexadecanol ≥99%

Synonyms: Cetyl alcohol, Palmityl alcohol

258741-100G 258741-5G 258741-500G
220049-127EA 46.09 USD
220049-127 220049-129 220049-128
1-Hexadecanol ≥99%
1-Hexadecanol
Formula: H₃C(CH₂)₁₄CH₂OH
MW: 242.45 g/mol
Boiling Pt: 305…330 °C (1013 hPa)
Melting Pt: 46…52 °C
Density: 0.81 g/cm³ (20 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004760
CAS Number: 36653-82-4
EINECS: 253-149-0
Merck Index: 13,02037

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR