4-Acetoxy-3-methoxybenzaldehyde ≥98%

258601-250G 258601-50G
220049-121EA 282.15 USD
220049-121 220049-122
4-Acetoxy-3-methoxybenzaldehyde ≥98%
4-Acetoxy-3-methoxybenzaldehyde
Formula: C₁₀H₁₀O₄
MDL Number: MFCD00003362
CAS Number: 881-68-5
EINECS: 212-920-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR