4-Ethoxyphenol ≥99%

Warning

258598-25G 258598-5G
220049-119EA 83.25 USD
220049-119 220049-120
4-Ethoxyphenol ≥99%
4-Ethoxyphenol
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 246 °C (1013 hPa)
Melting Pt: 63…64 °C
MDL Number: MFCD00002334
CAS Number: 622-62-8
EINECS: 210-748-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR