3,4-Dimethoxybenzoyl chloride ≥98%

Danger

Synonyms: Veratroyl chloride, 3,4-Dimethoxybenzene-1-carbonyl chloride

258040-5G 258040-25G
220049-074EA 59.34 USD
220049-074 220049-073
3,4-Dimethoxybenzoyl chloride ≥98%
3,4-Dimethoxybenzoyl chloride
Formula: C₉H₉ClO₃
MW: 200.62 g/mol
Boiling Pt: 95…98 °C (1 mmHg)
Melting Pt: 71 °C
MDL Number: MFCD00000674
CAS Number: 3535-37-3
EINECS: 222-568-0
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR