DL-α-Tocopherol

Synonyms: (±)-α-Tocopherol, Vitamin E Alcohol, all-rac-α-Tocopherol, 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, 2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-ol, DL-alpha-Tocopherol, Vitamin E

258024-100G 258024-5G 258024-500G
220049-070EA 151.2 USD
220049-070 220049-072 220049-071
DL-α-Tocopherol
DL-α-Tocopherol (Vitamin E)
Formula: C₂₉H₅₀O₂
MW: 430.71 g/mol
MDL Number: MFCD00072051
CAS Number: 10191-41-0
EINECS: 233-466-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR