2,5-Dimethoxybenzoic acid ≥98%

Warning

257966-25G 257966-5G
220049-066EA 570 USD
220049-066 220049-067
2,5-Dimethoxybenzoic acid ≥98%
2,5-Dimethoxybenzoic acid
Formula: C₉H₁₀O₄
MW: 182.18 g/mol
Boiling Pt: 314 °C (1013 hPa)
Melting Pt: 80 °C
MDL Number: MFCD00002436
CAS Number: 2785-98-0
EINECS: 220-503-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR