3-Methyl-1-butene ≥95%

Synonyms: Isoamylene, Isopropylethylene, Isopentene

257931-100G 257931-400G
220049-064EA 252 USD
220049-064 220049-065
3-Methyl-1-butene ≥95%
3-Methyl-1-butene
Formula: C₅H₁₀
MW: 70.13 g/mol
Boiling Pt: 20 °C (1013 hPa)
Melting Pt: -168 °C
Density: 0.6272 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00008937
CAS Number: 563-45-1
EINECS: 209-249-1
UN: 2561
ADR: 3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR