Selenium disulfide

Danger

Synonyms: Selenium sulphide, Selenium(IV) sulphide

257710-100G 257710-25G
220049-051EA 333.85 USD
220049-051 220049-052
Selenium disulfide
Selenium disulphide
Formula: S₂Se
MW: 143.09 g/mol
Boiling Pt: –112 °C (1013 hPa)
Density: 0.03 g/cm³ (-65 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011227
CAS Number: 7488-56-4
EINECS: 231-303-8
UN: 2657
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR