7-Amino-4-methylcoumarin

Synonyms: 7-Amino-4-methyl-2-benzopyrone, Coumarin 440, Coumarin 120, AMC

257370-100MG 257370-500MG
220048-008EA 73.35 USD
220048-008 220048-009
7-Amino-4-methylcoumarin
7-Amino-4-methylcoumarin
Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Boiling Pt: 352 °C (1013 hPa)
Melting Pt: 226 °C
Density: 1.377 g/cm³ (20 °C)
MDL Number: MFCD00006868
CAS Number: 26093-31-2
EINECS: 247-454-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR