Cadmium(II) telluride, deposition grade

Warning

Synonyms: Cadmium telluride

256544-10G 256544-50G
220048-938EA 124.62 USD
220048-938 220048-939
Cadmium(II) telluride, deposition grade
Cadmium(II) telluride
Formula: CdTe
MW: 240.01 g/mol
Melting Pt: 1089.85 °C
Density: 6.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00015998
CAS Number: 1306-25-8
EINECS: 215-149-9
UN: 2570
ADR: 6.1,III
Merck Index: 12,01667

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR